missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Hepatocyte Culture Media Kit, 500mL
BIOPHARMA

Product Code. 11533590 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11533590 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11533590 Supplier Corning™ Supplier No. 355056
  • ONLINE EXCLUSIVE PRICE
    282.00 €
    267.41 € / 1 sets
    Save 14.59 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Corning Hepatocyte Culture Medium (500 mL), EGF (5μg).

500 mL plus EGF supplement. This kit comprises a serum-free, fully-defined medium.

Corning Gentest cryopreserved animal hepatocytes allow researchers to study toxicological and pharmacokinetic properties of new chemical entities (NCEs) in vitro prior to expensive in vivo animal testing. Understanding species differences in metabolism is critical to selecting appropriate preclinical species for toxicity and pharmacokinetic studies. Corning Gentest animal cryohepatocytes are suitable for use in metabolic stability, metabolite profiling, and cytotoxicity studies.

  • Convenience – animal cryopreserved hepatocytes for rat, mouse, dog, and monkey are available as cryopreserved vialed suspension cells; just thaw and perform studies when needed
  • Full characterization – tested for high viability and phase I and II metabolic enzymes
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Differentiation Media
Sterility Sterile
For Use With (Application) Hepatocyte Culture
Quantity 1 Kit
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.