missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HYPERFlask™ M Cell Culture Vessels
BIOPHARMAGREENER_CHOICE

Product Code. 15554449 Shop All Corning Products
Click to view available options
Surface Coating:
CellBIND Treated
Not Treated
No. per Pack:
1/Pk.
4/Pk.
No. per Case:
24/Cs.
4/Cs.
Unit Size:
Pack of 24
Pack of 4
This item is not returnable. View return policy

Product Code. 15554449

Brand: Corning™ undefined

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.

The Corning High Yielding Performance Flask (HYPERFlask) cell culture vessel offers 1720 cm² growth area in the footprint of a traditional 175 cm² flask. This high yield, high performance flask utilizes a multilayered gas permeable growing surface for efficient gas exchange.

  • Innovative Design – 10 interconnected polystyrene growth surfaces
  • Optimal Growth – Corning CellBIND™ Surface treated gas permeable polystyrene for superior cell attachment and growth
  • Increase Cell Yield – 10-fold higher cell yields increases productivity and capacity and reduces processing time and incubator space
  • Save Time and Space – Reduce processing time and incubator storage space by handling one flask as compared to 10 traditional 175 cm² flasks
  • Automation Compatible – Bar coded and compatible
  • with The Automation Partnership SelecT™ and CompacT™ SelecT™ cell culture systems with an upgrade

The Corning HYPERFlask Cell Culture Vessel has the same external dimensions as a standard T 175 cm² flask with 10 times the cell yield.

Order Info

Supplied double bagged.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Yes
Closure Color Orange
Closure Material HDPE
Capacity (Metric) 560 mL
Material Polystyrene
Neck Style Straight
No. per Case 4/Cs.
No. per Pack 1/Pk.
Shape Rectangular
Surface Coating Not Treated
Graduated No
Sterility Sterile
Bottom Shape Rectangular
Closure Type Flat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.