missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Laminin, Mouse

Product Code. 10152421 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10152421 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152421 Supplier Corning™ Supplier No. 354232
  • ONLINE EXCLUSIVE PRICE
    245.71 €
    217.61 € / 1mg
    Save 28.10 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Promotes cell adhesion, migration, growth, and differentiation, including neurite outgrowth

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Laminin is a major component of basement membranes. It has numerous biological activities ­including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 to 10μg/ cm2
For Use With (Application) Cell Adhesion, Cell Growth and Differentiation, Cell Migration, Neurite Outgrowth
Formulation Frozen in 0.05M Tris HCl, 0.15M NaCl pH 7.4
Molecular Weight (g/mol) 900kD (unreduced)
Name Mouse Laminin
pH Range 7.4
Quality Control Testing >90% by SDS-PAGE
Quantity 1 mg
Source Engelbreth Holm Swarm (EHS) Mouse tumor
Storage Requirements -70°C
Cross Reactivity Mouse
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.