missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ LSE™ Swing-Out Rotor
Shop All Corning Products
Click to view available options
Capacity:
6 x 5 mL
Unit Size:
Each
Description
- For use with Corning LSE Compact Centrifuge (Cat. No. 07-201-417)
- 3500prm max.
Specifications
Specifications
| Type | Swing-out Rotor |
| Capacity | 6 x 5 mL |
| Max. Speed | 3,500 rpm |
| For Use With (Equipment) | Corning™ LSE™ Compact Centrifuge |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction