missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Microarray Slide Mailer Storage Boxes

Product Code. 10377672 Shop All Corning Products
Change view
Click to view available options
Clear 25 Slide Storage Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10377672 Clear 25 Slide Storage Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10377672 Supplier Corning™ Supplier No. 40083

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plastic containers function as storage boxes for printed arrays

  • Trans-orange plastic has low transmittance in the 500 to 700 nm wavelength range which helps protect Cy3 and Cy5 dyes from photobleaching
  • Rigid plastic containers do not shed particles or outgas volatile chemicals that may contaminate microarray slides

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50/Cs.
Color Orange
Type Storage Box
For Use With (Application) Printed microarrays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.