missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 2.5S Nerve Growth Factor (NGF), Mouse Natural

Product Code. 10584271 Shop All Corning Products
Click to view available options
Quantity:
1 mg
10 μ g
Unit Size:
1000µg
10µg
This item is not returnable. View return policy

Product Code. 10584271

Brand: Corning™ 356005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed specifically for use in basic cell biology research

  • NGFs are isolated from mouse submaxillary glands
  • 2.5S form consists of two β subunits
  • 7S form consists of two α, one β and two ϒ subunits
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma

Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.

Order Info

Both supplied in lyophilized form

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Growth Factor
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2%
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Purity or Quality Grade >90% by 4 to 20% Tris Glycine Gel Electrophoresis
Quantity 1 mg
Reactivity Mouse
Source Mouse Submaxillary Glands
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.