missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PET Storage Bottles, Square with Cap
BIOPHARMA

Product Code. 15118145 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Graduations:
100 to 1000 mL
100 to 500 mL
50 to 125 mL
50 to 250 mL
Unit Size:
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
15118145 1 L 100 to 1000 mL Pack of 24
15242596 125 mL 50 to 125 mL Pack of 24
15138125 250 mL 50 to 250 mL Pack of 24
15188135 500 mL 100 to 500 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15118145 Supplier Corning™ Supplier No. 431533
  • ONLINE EXCLUSIVE PRICE
    346.00 €
    323.01 € / Pack of 24
    Save 22.99 €
    6% Off
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

Features:

  • Good for storage of media, buffers and other aqueous solutions
  • Wide neck provides easier access for liquid transfer
  • Screened white enamel graduations are easier to read than molded graduations
  • Sterilized to SAL 10−6
  • Large white marking spot for easier identification and easy opening linear tear packaging
  • Individually bagged
  • Validated USP Class VI, noncytotoxic and nonpyrogenic


  •  
TRUSTED_SUSTAINABILITY

Specifications

Capacity 33.8 oz., 1000 mL
Capacity (Metric) 1 L
Color Orange
Diameter (English) 3.6 in.
Diameter (English) Inner Neck 1.4 in.
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
Material PET
No. per Case 24/Cs.
No. per Pack 12/Pk.
Quantity 12 Pk.
Sterility Sterile
Thread Size (Metric) GL 45
Closure Color Orange
Graduations 100 to 1000 mL
Closure Included Yes
Graduated Yes
Height (Metric) 216.5 mm
Height (English) 8.5 in.
Closure Material HDPE
Closure Size GL 45
Diameter (Metric) 92 mm
Closure Type Plug Seal
Autoclavable Not Autoclavable
Capacity (English) 33.8 oz.
Shape Square
Certifications/Compliance ISO 9001:2008
Disposable Single-use
Product Type Media Storage Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.