missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap
Shop All Corning Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Unit Size:
Pack of 10
Pack of 20
Pack of 6
Description
These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Color | Translucent |
| Material | Polypropylene |
| No. per Case | 10/Cs. |
| No. per Pack | 1/Pk. |
| Sterility | Non-sterile |
| Shape | Cylindrical |
| Height (Metric) | 20.4 cm |
| Diameter (Metric) | 10.8 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction