missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap
BIOPHARMA

Product Code. 10170213 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
Unit Size:
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10170213 1 L Pack of 24
10692434 150 mL Pack of 24
10160213 250 mL Pack of 24
10756323 500 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10170213 Supplier Corning™ Supplier No. 431433
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    195.79 € / Pack of 24
    Save 14.21 €
    6% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Linerless, one-piece, autoclavable orange GL45 threaded polypropylene plug-seal caps

Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.

Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.


Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.

  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

  • Screened white enamel graduations are easier to see than molded graduations

  • Sterilized by gamma radiation

  • Large white marking spot for easier identification

  • Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.

  • Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.

  • Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 30 oz., 1000 mL
    Capacity (Metric) 1 L
    Material Polycarbonate
    No. per Case 24/Cs.
    No. per Pack 12/Pk.
    Sterility Sterile
    Closure Color Orange
    Graduations 50 to 150 mL
    Closure Material Polypropylene
    Closure Size GL 45
    Closure Type Screw Cap
    Autoclavable Autoclavable
    Capacity (English) 30 oz.
    Shape Square
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.