missing translation for 'onlineSavingsMsg'
Learn More

Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat
GREENER_CHOICE

Product Code. 11513540 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11513540 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11513540 Supplier Corning™ Supplier No. 354115
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.

Order Info

Supplied in 100mL bottles; refrigerate as detailed on label

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Concentration 10 μg/mL
Product Type T-cell Culture Supplement
Quantity 100 mL
Source Rat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.