missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal Fit Pipet Tips: Bulk Packed
GREENER_CHOICE

Product Code. 10153761 Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Tip Style:
Beveled, Round
Round
Packaging:
1000 Tips/Bag, 1 Bag/Cs.
1000 Tips/Bag, 10 Bags/Cs.
1000 Tips/Bag, 10,000 Tips/Cs.
1000 Tips/Bag, 1000 Tips/Cs.
Unit Size:
Pack of 1000
Pack of 10000
This item is not returnable. View return policy

Product Code. 10153761

Brand: Corning™ 4866

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to assure a reliable fit with major brand pipetters

  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Yellow
Format Bulk
Tip Style Round
Packaging 1000 Tips/Bag, 1000 Tips/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.