missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Vented Screw Caps for Flasks, Sterile

Product Code. 10256821 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Falcon™ 175 cm2 flasks
Falcon™ 25 cm2 flasks
Falcon™ 75 cm2 flasks
Unit Size:
Pack of 100
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10256821 Falcon™ 25 cm2 flasks Pack of 100
10204861 Falcon™ 75 cm2 flasks Pack of 100
10709471 Falcon™ 175 cm2 flasks Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10256821 Supplier Falcon™ Supplier No. 354637
  • ONLINE EXCLUSIVE PRICE
    143.00 €
    135.60 € / Pack of 100
    Save 7.40 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with Falcon 25-, 75-, and 175cm2 flasks

  • Ensure consistent gas exchange and minimize contamination
  • Polyethylene caps with an integral, hydrophobic 0.2 μm microporous membrane filter vent allow consistent gas exchange when the caps are in the closed position.
  • Gases required or cell growth and metabolism pass freely through the vent while microorganisms cannot. Falcon vented caps will not wet out.
  • Vented caps minimize contamination associated with standard open incubation. The vented caps prevent media that can become trapped in a partially opened cap from blocking gas exchange. The caps will not fall off in the incubator because of vibration.
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polyethylene
Color Blue
For Use With (Equipment) Falcon™ 25 cm2 flasks
Includes 0.2μm Membrane vent
Product Type Cap, Vented
Sterility Sterile
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.