Learn More
Emma Safety Footwear Corona Safety Shoes
Product Code. 15928731
missing translation for 'orderingAttributeHoverText'
Size:
37
38
39
40
41
42
43
44
45
46
47
48
missing translation for 'unitSize'
1 pair
Product Code. 15928731
missing translation for 'mfr': Emma Safety Footwear MM54454044
Only null left
Add to basket
Add to basket
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. Highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist Outsole
  • Microfiber construction
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color White
Design Corona
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 44
Toe Material Steel