missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CorrectASE™ Enzyme, 50 Reactions

Product Code. 13442889
Change view
Click to view available options
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13442889 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13442889 Supplier Invitrogen™ Supplier No. A14972
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CorrectASE™ enzyme removes mismatches caused by oligonuceotide synthesis errors, leading to a 3–10 fold reduction in mutations in your synthetic genes or fragments.

CorrectASE™ enzyme removes mismatches caused by oligonuceotide synthesis errors, leading to a 3–10 fold reduction in mutations in your synthetic genes or fragments.

By introducing CorrectASE™ enzyme into your do-it-yourself gene syntheis workflow, you can:

  • Reduce the number of mutations in your synthetic gene or fragment
  • Reduce your labor time by screening only 2–4 clones instead of 10-16 clones per synthetic construct
  • Reduce your costs by sequencing only 2–4 clones instead of 10–16 synthetic genes

Prevent Unwanted Mutations
Commercially available synthetic oligonuceotides have a high error rate during synthesis, ranging from one per 300–1000 bases, depending on the source. These errors cause frameshift (deletion and insertion) and mismatch mutations during gene synthesis. Incubation with CorrectASE™ enzyme removes both type of mutations.

The incubation step with CorrectASE™ enzyme is introduced after the initial PCR assembly of oligonucleotides. The PCR product is denatured and reannealed so that any mutations will be unmatched. CorrectASE™ enzyme binds to the resulting mismatches and nicks both DNA strands 3' of the error. The 3' to 5' exonuclease activity of the enzyme removes the errors. A final PCR with a proofreading polymerase then assembles the corrected fragments, thus increasing the likelihood of isolating clones with the correct sequence. Depending upon the incoming oligonuceotide quality, only 2–4 clones need to be screened, compared to 10–16 clones in a workflow that does not include the correction step. Including CorrectASE™ enzyme in your gene synthesis workflow decreases labor time and sequencing costs.

TRUSTED_SUSTAINABILITY

Specifications

Purity >95% by SDS-PAGE
Content And Storage • CorrectASE, 1 tube (50 reactions)
• 10X CorrectASE Reaction Buffer, 1 tube
Exonuclease Activity 3'–5'
Enzyme CorrectASE™
Compatible Buffer Reaction Buffer
Quantity 50 Reactions
Product Type CorrectASE™ Enzyme

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.