missing translation for 'onlineSavingsMsg'
Learn More

Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals

Product Code. 13581640 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 13581640

Brand: Thermo Scientific Acros 448960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-04-4
Molecular Formula C23H30O6
Molecular Weight (g/mol) 402.49
MDL Number MFCD00003609
InChI Key ITRJWOMZKQRYTA-RFZYENFJSA-N
Synonym cortisone acetate, cortisone 21-acetate, cortone acetate, incortin, biocort acetate, artriona, cortadren, scheroson, cortisyl, adreson
PubChem CID 5745
ChEBI CHEBI:3897
IUPAC Name [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

Specifications

Melting Point 237°C to 240°C
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,2525
Formula Weight 402.48
Percent Purity 98+%
Chemical Name or Material Cortisone 21-acetate
compliance-icons
Product Identifier
  • Cortisone 21-acetate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.