missing translation for 'onlineSavingsMsg'
Learn More

Cortisone, TRC

Product Code. 17963295
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17963295 1 g 1g
17954835 25 g 25g
17911925 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963295 Supplier Toronto Research Chemicals Supplier No. TRCC6965001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-06-5
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Cortisone (8CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione, 11-Dehydro-17-hydroxycorticosterone, 17-Hydroxy-11-dehydrocorticosterone, 17α,21-Dihydroxy-4-pregnene-3,11,20-trione, 17α,21β-Dihydroxy-4-pregnene-3,11,20-trione, 17α-Hydroxy-11-dehydrocorticosterone, Adrenalex, Compound E, Cortisate, Cortivite, Cortogen, Cortone, KE, Kendall's compound E, NSC 9703, Pregn-4-en-17α,21-diol-3,11,20-trione, Reichstein's substance Fa, Wintersteiner's compound F, δ4-Pregnene-17α,21-diol-3,11,20-trione
IUPAC Name (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1
Formula Weight 360.1937
Percent Purity >95%
Chemical Name or Material Cortisone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.