missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Cluster Tube System

Product Code. 10256531 Shop All Corning Products
Change view
Click to view available options
Product Type:
8-Strip Microplate Cluster Tube
Microplate Cluster Tube
Format:
Strip
Tube
Quantity:
12 Strips/Rack, 120 Strips/Cs.
12/Rack
96/Rack
Bulk
Bulk, 120/Bag
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 120
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Format Quantity Sterility Unit Size
10256531 Microplate Cluster Tube Tube Bulk Non-sterile Pack of 960
11763994 8-Strip Microplate Cluster Tube Strip 12 Strips/Rack, 120 Strips/Cs. Non-sterile Pack of 120
10681432 8-Strip Microplate Cluster Tube Strip 12/Rack Sterile Pack of 120
10085431 8-Strip Microplate Cluster Tube Strip Bulk, 120/Bag Non-sterile Pack of 120
10285471 Microplate Cluster Tube Tube 96/Rack Non-sterile Pack of 960
10690112 Microplate Cluster Tube Tube 96/Rack Sterile Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10256531 Supplier Corning™ Supplier No. 4401
  • ONLINE EXCLUSIVE PRICE
    125.00 €
    115.99 € / Pack of 960
    Save 9.01 €
    7% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.

  • Composed of 96 polypropylene tubes in a standard microplate format.
  • Features 1.2 mL tubes that are available individually or in strips of eight tubes.
  • Polyethylene tube caps are available in 8-cap strips.
TRUSTED_SUSTAINABILITY

Specifications

Format Tube
Quantity Bulk
Autoclavable Autoclavable
Material Polypropylene
Product Type Microplate Cluster Tube
Sterility Non-sterile
No. per Pack 960/Pk.
No. per Case 960/Cs.
Color Translucent
Volume (Metric) 1 mL
For Use With (Equipment) Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges
Volume (English) 0.033 oz.
Well Count 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.