missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles
BIOPHARMA

Product Code. 10539082 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Unit Size:
Pack of 12
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10539082 250 mL Pack of 12
10052491 125 mL Pack of 24
10179200 500 mL Pack of 12
10177500 1 L Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10539082 Supplier Corning™ Supplier No. 8390
  • ONLINE EXCLUSIVE PRICE
    100.48 €
    90.20 € / Pack of 12
    Save 10.28 €
    10% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions

  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Costar™ traditional style has smooth sides
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.45 oz., 250 mL
Capacity (Metric) 250 mL
Color Clear
Material Polystyrene
No. per Case 12/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Closure Color Red
Neck Finish 45
Closure Included Yes
Graduated Yes
Mouth Wide, Threaded
Closure Material HDPE
Closure Size 45 mm
Closure Type Plug Seal
Autoclavable Not Autoclavable
Capacity (English) 8.45 oz.
Shape Cylindrical
Disposable Single-use
Product Type Storage Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.