Learn More
Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Product Code. 10718402 Shop All Corning Products
Change view
Click to view available options
Color:
Assorted
Natural
Capacity (Metric):
0.65 mL
1.7 mL
2 mL
Packaging:
1000/Pk.
500/Pk.
5000/Pk.
Unit Size:
Pack of 1000
Pack of 500
Pack of 5000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (Metric) Packaging unitSize
10718402 Natural 0.65 mL 1000/Pk. Pack of 1000
10611442 Assorted 0.65 mL 1000/Pk. Pack of 1000
10276141 Natural 2 mL 1000/Pk. Pack of 1000
10429521 Natural 1.7 mL 500/Pk. Pack of 500
10467221 Assorted 1.7 mL 500/Pk. Pack of 500
10439521 Natural 1.7 mL 5000/Pk. Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10718402 Supplier Corning™ Supplier No. 3208
Add to basket
Add to basket
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling.
  • Withstands a maximum RCF of 17,000 x g.
  • RNase-/DNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Snap Cap
Certifications/Compliance RNase, DNase-free
Closure Color Natural
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 0.65 mL
Color Natural
Material Polypropylene
No. per Pack 500/Pk.
Packaging 1000/Pk.
Sterility Non-sterile
Graduated Yes
Closure Type Snap Cap
Bottom Shape Conical
Max. RCF 17,000 x g
Product Type Microcentrifuge Tube