missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
BIOPHARMAGREENER_CHOICE

Product Code. p-176666 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.4 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Diameter (Metric) Outer:
6.5 mm
12 mm
24.5 mm
Culture Area:
0.33 cm2
1.12 cm2
4.7 cm2
Unit Size:
Pack of 24
Pack of 48
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size No. of Wells Diameter (Metric) Outer Culture Area unitSize
10301031 0.4 μm 6 24.5 mm 4.7 cm2 Pack of 24
10567522 0.4 μm 12 12 mm 1.12 cm2 Pack of 48
10615345 3 μm 12 12 mm 1.12 cm2 Pack of 48
10246481 8 μm 6 24.5 mm 4.7 cm2 Pack of 24
10025471 3 μm 24 6.5 mm 0.33 cm2 Pack of 48
10107341 5 μm 24 6.5 mm 0.33 cm2 Pack of 48
10147291 0.4 μm 24 6.5 mm 0.33 cm2 Pack of 48
10167000 8 μm 24 6.5 mm 0.33 cm2 Pack of 48
10471242 3 μm 6 24.5 mm 4.7 cm2 Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10301031 Supplier Corning™ Supplier No. 3412
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Description 6 Inserts; 4 plates/Cs.
Culture Area 4.7 cm2
Material Polycarbonate
Volume (Metric) Well 2.6 mL
Pore Size 0.4 μm
Type Pre-loaded Microporous Insert in Tissue Culture Plate
For Use With (Application) Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
Diameter (Metric) Outer 24.5 mm
Quantity Packaged 6 inserts in a 6-well plate, 4 plates per case
No. per Pack 4 plates; 6 inserts/plate
Surface Coating TC-Treated
Color Clear
Surface Treatment TC-Treated
Sterility Sterile
No. of Wells 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.