missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cotinine, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Cotinine ((-)-Cotinine), an alkaloid in tobacco and a major metabolite of nicotine, is used as a biological indicator to measure the composition of tobacco smoke
Specifications
Specifications
| Chemical Name or Material | Cotinine |
| CAS | 486-56-6 |
| Color | Light Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C10H12N2O |
| Quantity | 50 mg |
| Synonym | (-)-Cotinine (S)-Cotinine NIH-10498 |
| Solubility Information | Ethanol : 120 mg/mL (680.97 mM; Need ultrasonic) ∣DMSO : 65 mg/mL (368.86 mM; Need ultrasonic) |
| SMILES | O=C1N(C)[C@H](C2=CC=CN=C2)CC1 |
| Molecular Weight (g/mol) | 176.22 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction