missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Excellent Interfolded 2 Ply Paper Hand Towels 6604, 20 Packs

Product Code. 11728188 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Thickness: 2-ply; White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11728188 Thickness: 2-ply; White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11728188 Supplier Kimberly-Clark™ Supplier No. 6604

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hand towels in a clever interfolded design prevents ripping and dispenses single towels each time. Airflex technology ensures each cover is absorbent even when wet.

20 x 190 white, 2 ply sheets (3,800 total) 20 x 190 white, 2 ply sheets (3,800 total) Clever interfolded design to prevent ripping and ensuring only single towels are dispensed each time. Soft, strong and highly absorbent, even when wet. 100% recycled fibre and carries the European Ecolabel Compatible with Aquarius Folded Hand Towel Dispensers (product codes 6954 and 6956) Scott Excellent Interfolded Hand Towels are softer, more absorbent and Stronger than conventional towels, providing consistent quality, at an affordable price. The clever interfolded design helps to prevent towels from ripping when being pulled from the dispenser, and ensures only single towels are dispensed each time, reducing mess and wastage. These hand towels are made from 100% recycled fibre and carry the European Ecolabel, your guarantee that they are manufactured responsibly at every step of the process. Individual sheet size 23 cm (L) x 25 cm (W).Compatible with Aquarius Folded Hand Towel Dispensers (product codes 6954 and 6956). 20 x 190 white, 2 ply sheets (3,800 total)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Towel
Color White
Material Paper
Sterility Non-sterile
For Use With (Application) Nurse Stations, Food Contact Areas, Busy Washrooms in Hospitals, Public Venues, Office Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.