missing translation for 'onlineSavingsMsg'
Learn More

Coumachlor, TRC

Product Code. 30367426
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367426 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367426 Supplier Toronto Research Chemicals Supplier No. TRCC7450001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coumachlor
CAS 81-82-3
Molecular Formula C19 H15 Cl O4
Quantity 1 g
Synonym Coumachlor (6CI), Coumarin, 3-(α-acetonyl-p-chlorobenzyl)-4-hydroxy- (7CI,8CI), 3-[1-(4-Chlorophenyl)-3-oxobutyl]-4-hydroxy-2H-1-benzopyran-2-one, (±)-3-(α-Acetonyl-4-chlorobenzyl)-4-hydroxy coumarin, (±)-Coumachlor, (±)-p-Chlorowarfarin, 3-(α-Acetonyl-4-chlorobenzyl)-4-hydroxycoumarin, 3-(α-p-Chlorophenyl-β-acetylethyl)-4-hydroxycoumarin, 3-[1-(p-Chlorophenyl)-2-acetylethyl]-4-hydroxycoumarin, Cumachlor, DL-3-(α-Acetonyl-4-chlorobenzyl)-4-hydroxycoumarin, Experimental Rodenticide 332, Geigy Rodenticide Exp. 332, Racemic coumachlor, Tomorin, p-Chlorowarfarin
InChI Formula InChI=1S/C19H15ClO4/c1-11(21)10-15(12-6-8-13(20)9-7-12)17-18(22)14-4-2-3-5-16(14)24-19(17)23/h2-9,15,22H,10H2,1H3
SMILES CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccc(Cl)cc3
IUPAC Name 3-[1-(4-chlorophenyl)-3-oxobutyl]-4-hydroxychromen-2-one
Molecular Weight (g/mol) 342.77
Formula Weight 342.0659
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.