missing translation for 'onlineSavingsMsg'
Learn More

Coumalic acid, 97%

Product Code. 11497196
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497196 5 g 5g
11407206 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497196 Supplier Thermo Scientific Alfa Aesar Supplier No. B25521.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 500-05-0 | C6H4O4 | 140.09 g/mol

Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-05-0
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00006644
InChI Key ORGPJDKNYMVLFL-UHFFFAOYSA-N
Synonym coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g
PubChem CID 68141
SMILES OC(=O)C1=COC(=O)C=C1

Specifications

Melting Point ∼208°C (decomposition)
Boiling Point 218°C (120 mmHg)
Quantity 5 g
Beilstein 119747
Merck Index 14,2557
Solubility Information Very soluble in water.
IUPAC Name 6-oxopyran-3-carboxylic acid
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material Coumalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.