missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-3-carboxylic acid, TRC

Product Code. 30357238
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357238 Supplier Toronto Research Chemicals Supplier No. TRCC7555285G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coumarin-3-carboxylic acid
CAS 531-81-7
Molecular Formula C10H6O4
Quantity 5 g
Synonym 2-Oxo-2H-1-benzopyran-3-carboxylic Acid
InChI Formula InChI=1S/C10H6O4/c11-9(12)7-5-6-3-1-2-4-8(6)14-10(7)13/h1-5H,(H,11,12)
SMILES OC(=O)C1=Cc2ccccc2OC1=O
IUPAC Name 2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol) 190.15
Formula Weight 190.0266
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.