missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Countess™ Cell Counting Chamber Slides

Product Code. 12098489
Change view
Click to view available options
Type:
Cell Counting Chamber Slide
Reusable Slide
Includes:
No Stain
SafeCount Stain, 2 x 1 mL Vials
Trypan Blue Stain, 100 mL Bottle
Trypan Blue Stain, 2 x 1 mL Vials
Trypan Blue Stain, 2 x 100 mL Bottles
Trypan Blue Stain, 20 x 1 mL Vials
Quantity:
1 Glass Slide
1250 Slides
2500 Slides
50 Slides
500 Slides
5000 Slides
Unit Size:
1 set
Each
Pack of 2500
Pack of 50
Pack of 500
Pack of 5000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Includes Quantity Unit Size
12098489 Cell Counting Chamber Slide Trypan Blue Stain, 20 x 1 mL Vials 500 Slides Pack of 500
15311986 Reusable Slide No Stain 1 Glass Slide Each
31781208 Cell Counting Chamber Slide SafeCount Stain, 2 x 1 mL Vials 50 Slides 1 set
11544596 Cell Counting Chamber Slide No Stain 50 Slides Each
10399053 Cell Counting Chamber Slide Trypan Blue Stain, 2 x 1 mL Vials 50 Slides Pack of 50
12157220 Cell Counting Chamber Slide Trypan Blue Stain, 100 mL Bottle 1250 Slides Each
12167220 Cell Counting Chamber Slide Trypan Blue Stain, 100 mL Bottle 2500 Slides Pack of 2500
10665865 Cell Counting Chamber Slide Trypan Blue Stain, 2 x 100 mL Bottles 5000 Slides Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12098489 Supplier Invitrogen™ Supplier No. C10312
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Countess Cell Counting Chamber Slides are conveniently packaged for use with the Countess Automated Cell Counters. Each disposable and reusable slide features two enclosed chambers.

The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.

The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.

The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.

Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.

The Countess Automated Cell Counter works in three simple steps:

  • Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
  • Insert the slide into the instrument
  • Press 'Count'

That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.

TRUSTED_SUSTAINABILITY

Specifications

Type Cell Counting Chamber Slide
Product Line Countess
For Use With (Equipment) Countess Automated Cell Counter
Holds 10 μL/Chamber
Includes Trypan Blue Stain, 20 x 1 mL Vials
Positions 2 Chambers/Slide
Depth (Metric) 100 μm (Chamber)
Dimensions 75 x 25 x 1.8 mm (Chamber)
Content And Storage • Countess™ Cell Counting Chamber Slides, 10 x 50 slides
• Trypan blue stain, 20 x 1 mL vials

Each slide contains two cell counting chambers.
For use at room temperature under normal laboratory conditions.
Quantity 500 Slides
Do you offer an alternative for Countess Cell Counting Chamber Slides (Cat. no. C10283) for Countess 3 and Countess 3 FL Automated Cell Counter?

Yes. The alternative options for Countess Cell Counting Chamber Slides (Cat. No. C10283) and for Countess 3 and Countess 3 FL Automated Cell Counter can be found using the link below:

Countess Cell Counting Chamber Slides (Cat. Nos. C10228, C10312, C10313, C10314, C10315)

What Countess Automated Cell Counter instruments are compatible with the Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315)?

The Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315) are compatible with all Countess Automated Cell Counter instruments.

compliance-icons
Product Identifier
  • chamber slides countess 1000 counts
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,7-Naphthalenedisulfonic acid, 3,3-[(3,3-dimethyl[1,1-biphenyl]-4,4-diy l)bis(azo)]bis[5-amino-4-hy

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.