Learn More
Invitrogen™ Countess™ Cell Counting Chamber Slides
Descrizione
The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.
The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.
The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.
Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.
The Countess Automated Cell Counter works in three simple steps:
- Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
- Insert the slide into the instrument
- Press 'Count'
That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.
Specifica
Specifica
| Type | Cell Counting Chamber Slide |
| For Use With (Equipment) | Countess 3/3FL Automated Cell Counters |
| Coupler Option | Dead Cell Stain |
| Includes | SafeCount Stain, 2 x 1 mL Vials |
| Content And Storage | 50 Pk. Chamber Slides and SafeCount (Cat. No. A40008024), Store at Room Temp |
| Quantity | 50 Slides |
For Research Use Only. Not for use in diagnostic procedures.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.