Learn More
Hecht Karl™ Counting Chamber
Product Code. 10647564
Change view
Click to view available options
Grid Type:
16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm
Unit Size:
Each
Product Code. Grid Type Unit Size
10647564 16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm Each
1 options
Product Code. 10647564 Supplier Hecht Karl™ Supplier No. 40449712
  • ONLINE EXCLUSIVE PRICE
    118.53 €
    69.05 € / Pack of 1
    Save 49.48 €
    41% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Fuchs-Rosenthal Counting Chamber
Grid Type 16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm
For Use With (Application) Commonly used to count cells in cerebrospinal fluid