Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Product Code. 10638902
Change view
Click to view available options
Description:
Counting chamber acc. to Bürker
Counting chamber acc. to Nageotte
Counting chamber acc. to Thoma
Counting chamber acc. to Thoma - new
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10638902 Counting chamber acc. to Thoma Each
10448851 Counting chamber acc. to Thoma - new Each
10521412 Counting chamber acc. to Nageotte Each
10438701 Counting chamber acc. to Bürker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10638902 Supplier Hirschmann™ Supplier No. 8100105
  • ONLINE EXCLUSIVE PRICE
    71.00 €
    63.00 € / Pack of 1
    Save 8.00 €
    11% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
  • Double net ruling
  • Including 2 cover glasses for hemacytometer
  • CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Product Type Counting chambers, double net ruling
Description Counting chamber acc. to Thoma
Quantity 1 Each
Depth (Metric) 0.1 mm
Includes 2 cover glasses for haemacytometer