Learn More
Hirschmann™ Cover Glasses for Microscopy
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
Brand: Hirschmann™ 8000117
Code : 04
Additional Details : Weight : 0.25000kg
Description
- 0.13-0.17 mm thickness
- Acc. to DIN 58 884, CE acc. IVD 98/79 EC
Specifications
Coverslip | |
24 x 46 mm | |
10 x 100/Pk. |
#1 (0.13 to 0.16 mm) | |
DIN 58 884, CE acc. IVD 98/79 EC |