Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
Product Code. 15340541 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15340541 X-Large Each
15310541 Small Each
15320541 Medium Each
15330541 Large Each
15360541 3X-Large Each
15350541 2X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15340541 Supplier DuPont™ Supplier No. D14886064
  • ONLINE EXCLUSIVE PRICE
    6.77 €
    5.40 € / Pack of 1
    Save 1.37 €
    20% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
  • Robust yet lightweight
  • Elasticated face, wrists, waist and ankles
  • Antistatic treatment on the outside
  • Stitched external seams in yellow for visual identification and differentiation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size X-Large
Material Polyethylene
Color White
Ankle Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type Zipper
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-1
Size (Metric) Chest 108 to 116 cm