missing translation for 'onlineSavingsMsg'
Learn More

Bollé Safety Coverall Clear Safety Goggles

Product Code. 15548281 Shop All Bollé Safety Products
Change view
Click to view available options
Model:
Coverall Autoclave
Coverall Clean
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15548281 Coverall Clean Pack of 5
12697723 Coverall Autoclave Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15548281 Supplier Bollé Safety Supplier No. COVACLEAN
  • ONLINE EXCLUSIVE PRICE
    41.95 €
    38.17 € / Pack of 5
    Save 3.78 €
    9% Off
View all promotions
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protective goggles is specially designed to guarantee total comfort and optimum protection in controlled environments or sensitive and sterile environments such as pharmacies, micro-electronics, and operating theatres.

Coverall Autoclave:

  • High fogging resistance
  • Autoclave compatible product
  • Overflow chute for liquids
  • PLATINUM coating : anti-scratch & anti-fog

Coverall Clean:

  • Can be sterilized using gamma, beta irrdiation or etylene oxide sterilization
  • Disposable
  • Overflow chute for liquids
  • PLATINUM LITE, anti-fog coating
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Pharmaceutics, micro-electronics and surgery departments
Includes Neoprene strap
Model Coverall Clean
Weight (Metric) 79 g
Material Polycarbonate
Lens Material Polycarbonate
Frame Material PVC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.