missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Covers with Ring Handle, Translucent

Product Code. p-7156863
Change view
Click to view available options
For Use With (Equipment):
CR 15 Crucible
CR 2 Crucible
CR 4 Crucible
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15858064 CR 2 Crucible Each
15868064 CR 15 Crucible Each
15878064 CR 4 Crucible Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15858064 Supplier Helios Italquartz Supplier No. 64F00502
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Covers with ring handle constructed of translucent, durable, resistant, and chemically inert quartz glass.

  • Translucent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass

Specifications

Color Translucent
Material Quartz
Type Covers with Ring Handle
For Use With (Equipment) CR 2 Crucible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.