missing translation for 'onlineSavingsMsg'
Learn More

Ibidi Coverslips for Sticky-Slides

Product Code. 15708052
Encompass_Preferred
Click to view available options
:
Coverslips
This item is not returnable. View return policy

Product Code. 15708052

Brand: Ibidi 10812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Coverslips for Sticky-Slides are coverslips #1.5H (170 μm +/- 5 μm) D 263 M Schott glass.

  • Coverslips #1.5H (170 μm ± 5 μm) D 263 M Schott glass
  • 25 mm x 75 mm
  • non-sterile

Specifications

Product Type Coverslip
Length (Metric) 75 mm
Width (Metric) 25 mm
Thickness (Metric) 170 μm
Quantity 100
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.