Learn More
SureScreen Diagnostics COVID-19 Visual Antigen Rapid Test Cassette
COVID-19 Visual Antigen Rapid Test Cassette using nasal (anterior), nasopharyngeal and oropharyngeal swab samples.
Brand:  SureScreen Diagnostics COVID19AGVC
Additional Details : Weight : 0.40000kg
Product Code. 17033987
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alert:
For professional in vitro diagnostic use only.
This product is CE-IVD marked for professional use and complies with the European In-Vitro Diagnostic Devices Directive (98/79/EC).
Description
Description
  • For the direct and qualtitative detection of SARS-CoV-2 viral nucleoprotein antigens from nasal (anterior), nasopharyngeal and oropharyngeal swab samples for individuals suspected of COVID-19
  • No additional equipment required
Specifications
Specifications
Yes
95%
25 Tests/Box
15 min.
Nose and Throat Swab
Viral Rapid Test
100%