Learn More
Sperian™ Cowhide Leather Gloves
Product Code. 12625553
Change view
Click to view available options
Size: 9
Product Code.
12625553 Size: 9
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12625553 Supplier Sperian™ Supplier No. 228156109
  • ONLINE EXCLUSIVE PRICE
    58.05 €
    32.06 € / Pack of 1
    Save 25.99 €
    44% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Color Yellow
Material Cowhide Leather
Product Type Gloves
Size 9
Thickness (Metric) 1.3 mm
Model T09
Resistant To Thermal, Cuts