missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CP 154526

Product Code. 18095012
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095012 50 mg 50mg
18074272 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095012 Supplier R&D Systems™ Supplier No. 2779/50
  • ONLINE EXCLUSIVE PRICE
    1294.23 €
    1165.50 € / 50mg
    Save 128.73 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective, non-peptide CRF1 antagonist

Selective, non-peptide CRF1 receptor antagonist (Ki values are 2.7 and > 10000 nM for CRF1 and CRF2 receptors respectively). Blocks CRF-induced adenylate cyclase activation (Ki = 3.7 nM) and attenuates activation of the HPA axis by CRF. Displays anxiolytic-like activity in the rat elevated plus-maze test; brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 257639-98-8
Molecular Formula C23H33ClN4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 400.99
Purity >99%
Molecular Weight (g/mol) 400.995
Chemical Name or Material CP 154526
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.