missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CP 690550 citrate

Product Code. 30001531
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001531 50 mg 50mg
30001514 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001531 Supplier R&D Systems™ Supplier No. 4556/50
  • ONLINE EXCLUSIVE PRICE
    1217.50 €
    1092.00 € / 50mg
    Save 125.50 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent JAK inhibitor

Potent JAK inhibitor (IC50 values are 1, 20, and 112 nM for JAK3, JAK2 and JAK1 respectively). Orally active immunosuppressant; exhibits efficacy in rodent rheumatoid arthritis models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540737-29-9
Molecular Formula C22H28N6O8

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 504.49
Purity >99%
Molecular Weight (g/mol) 504.5
Chemical Name or Material CP 690550 citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.