missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CP 93129 dihydrochloride

Product Code. 30001168
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001168 50 mg 50mg
18064042 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001168 Supplier R&D Systems™ Supplier No. 1032/50
  • ONLINE EXCLUSIVE PRICE
    1074.26 €
    955.50 € / 50mg
    Save 118.76 €
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT1B agonist

Potent and highly selective 5-HT1B agonist (Ki values are 8.1, 1100, 1500, 2900 and 7200 nM for 5-HT1B, 5-HT1D, 5-HT1A, 5-HT1c and 5-HT2, respectively). Reduces food intake and decreases body weight in rats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879089-64-2
Molecular Formula C12H17Cl2N3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 288.18
Purity >98%
Molecular Weight (g/mol) 306.187
Chemical Name or Material CP 93129 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.