Learn More
Agilent Technologies CP-Al2O3/KCl, 50 m, 0.53 mm, 10.00 μm, 7 inchCP-Al2O3/KCl, 50 m, 0.53 mm, 10.00 μm, 7 inch
Brand:  Agilent Technologies CP7518
Encompass_Preferred
CP-Al2O3/KCl, 50 m, 0.53 mm, 10.00 μm, 7 inchCP-Al2O3/KCl, 50 m, 0.53 mm, 10.00 μm, 7 inch
Product Code. 15507280
  • 2216.70 € / Pack of 1
Only null left
Estimated Shipment: 23-04-2026