missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Plastic Quick-Disconnect Fittings

Product Code. 11749979
Change view
Click to view available options
Diameter (Metric) Inner:
6.4 mm
9.5 mm
Product Type:
Hose Barb Non-valved Coupling Insert
Hose Barb Non-valved In-line Coupling Body
Hose Barb Non-valved In-line Coupling Insert
Material:
ABS
Polycarbonate
Polysulfone
Color:
Amber
Purple
White
Unit Size:
Pack of 250
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Product Type Material Color unitSize
11749979 9.5 mm Hose Barb Non-valved In-line Coupling Insert Polycarbonate Amber Pack of 5
17385589 9.5 mm Hose Barb Non-valved In-line Coupling Insert ABS White Pack of 250
17729529 6.4 mm Hose Barb Non-valved In-line Coupling Body Polycarbonate Purple Pack of 250
11780029 6.4 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber Pack of 5
11739979 6.4 mm Hose Barb Non-valved Coupling Insert Polysulfone Amber Pack of 5
17739529 6.4 mm Hose Barb Non-valved In-line Coupling Insert Polycarbonate Purple Pack of 250
11700629 9.5 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11749979 Supplier CPC™ Supplier No. 3131172

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifications

Certifications/Compliance USP Class VI, ADCF
Autoclavable Autoclavable
Diameter (English) Inner 0.37 in.
Diameter (Metric) Inner 9.5 mm
Material Polycarbonate
Overall Length (English) 1.3 in.
Overall Length (Metric) 33 mm
For Use With 9.5mm
For Use With (Application) Cell culture media packaging and transfer lines, harvest and collection applications
Min. Operating Temperature (English) -40°F
Min. Operating Temperature (Metric) -40°C
Height (Metric) 15.2 mm
Height (English) 0.6 in.
Material (Seal) Platinum-cured Silicone
Color Amber
Maximum Pressure 60 psi
Max. Operating Temperature (English) 300°F
Max. Operating Temperature (Metric) 149°C
Product Type Hose Barb Non-valved In-line Coupling Insert
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.