Learn More
Chemsplash CPE OverShoe Compressed Polyethylene -Machine Made
GREENER_CHOICE
Product Code. 30058405
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30058405 Blue Pack of 2000
30058232 White Pack of 2000
30058460 Red Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30058405 Supplier Chemsplash Supplier No. 2522.300.999
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Elasticated Overshoe in a 3 g. 16 in. compressed polyethylene material.
Elasticated Overshoe
  • 3 GSM Compressed Polyethylene Material
  • 16 in. Long
  • Elasticated Ankles
  • Heat Sealed Seams
  • PFAS Free
Description: Elasticated Overshoe in a 3 gram blue compressed polyethylene material. Creates a barrier over your footwear. The 16 in. length offers suitability for all footwear sizing. Ideal for keeping clean footwear clean or keeping clean floors clean from dirty footwear.
Specifications
Size 3 g
Material Compressed Polyethylene
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Plain
Resistance Liquid
Product Type Shoe Covers