missing translation for 'onlineSavingsMsg'
Learn More

CPI-444, MedChemExpress

Product Code. 16484746
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484746 100 mg 100mg
16480057 50 mg 50mg
16463668 10 mg 10mg
16436407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484746 Supplier Medchem Express Supplier No. HY101978
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CPI-444 (V81444) is a potent, orally active and selective adenosine A2A receptor (A2AR) antagonist, which induces antitumor responses.

CPI-444 (V81444) is a potent, orally active and selective adenosine A2A receptor (A2AR) antagonist, which induces antitumor responses.

Specifications

Chemical Name or Material CPI-444
CAS 1202402-40-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H21N7O3
Quantity 100 mg
Synonym V81444 ciforadenant
Solubility Information DMSO : 66.67 mg/mL (163.64 mM; Need ultrasonic)
SMILES NC1=NC(C2=CC=C(C)O2)=C(N=NN3CC4=NC(CO[C@@H]5COCC5)=CC=C4)C3=N1
Molecular Weight (g/mol) 407.43
Formula Weight 407.43
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.