missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CPT 11

Product Code. 18752534
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752534 10 mg 10mg
18712624 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752534 Supplier R&D Systems™ Supplier No. 2688/10
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    239.40 € / 10mg
    Save 26.60 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA topoisomerase I inhibitor; antitumor

Inhibitor of DNA topoisomerase I that displays antitumor activity against a range of tumor types. Active metabolite available ( SN 38, Cat. No. 2684).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100286-90-6
Molecular Formula C33H39ClN4O6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 623.14
Purity >99%
Molecular Weight (g/mol) 623.15
Chemical Name or Material CPT 11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.