missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CPYPP

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CPYPP is a DOCK2-Rac1 interaction inhibitor; suppresses DOCK2-induced Rac1 activation. Also inhibits DOCK1 and DOCK5. Exhibits selectivity over DOCK9. Reduces chemotactic responses in vitro and in vivo, and activates T cells in vitro. Also attenuates HER2-mediated breast cancer cell migration in vitro.
Specifications
Specifications
| Chemical Name or Material | Rho GTPase Inhibitors |
| CAS | 310460-39-0 |
| Quantity | 10 mg |
| Target | Small G Protein Inhibitors |
| Molecular Formula | C18H13ClN2O2 |
| Formula Weight | 324.76 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction