missing translation for 'onlineSavingsMsg'
Learn More

Creatine, anhydrous, 98%

Product Code. 11488077
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488077 100 g 100g
11478077 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488077 Supplier Thermo Scientific Alfa Aesar Supplier No. A17477.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 57-00-1 | C4H9N3O2 | 131.135 g/mol

Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-00-1
Molecular Formula C4H9N3O2
Molecular Weight (g/mol) 131.135
MDL Number MFCD00004282
InChI Key CVSVTCORWBXHQV-UHFFFAOYSA-N
Synonym creatine, creatin, kreatin, krebiozon, n-amidinosarcosine, n-methyl-n-guanylglycine, methylglycocyamine, 2-1-methylguanidino acetic acid, creatine, hydrate, alpha-methylguanido acetic acid
PubChem CID 586
ChEBI CHEBI:16919
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=N)N

Specifications

Melting Point ∽290°C (decomposition)
Density 1.33
Assay Percent Range 98%
Quantity 100 g
Beilstein 907175
Merck Index 14,2568
Solubility Information Soluble in water.
Formula Weight 131.13
Percent Purity 98%
Chemical Name or Material Creatine, Anhydrous, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.