missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine-d3 H2O (methyl-d3), CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | Creatine-(methyl-d3) Monohydrate |
| CAS | 284664-86-4 |
| Molecular Formula | C4 D3 H6 N3 O2 . H2 O |
| Quantity | 0.05 g |
| Synonym | Glycine, N-(aminoiminomethyl)-N-(methyl-d3)-, hydrate (1:1) (ACI), Glycine, N-(aminoiminomethyl)-N-(methyl-d3)-, monohydrate (9 CI), N-(Aminoiminomethyl)-N-(methyl-d3)glycine monohydrate, Creatine-d3 monohydrate, Creatine-D3 (N-methyl-D3) monohydrate, 2-(1-Trideuteriomethylguanidino)acetic acid monohydrate, N-(Methyl-d3)-N-guanylglycine monohydrate, [N-(Amidino)-N-(methyl-d3)amino]acetic acid monohydrate |
| InChI Formula | InChI=1 S/C4H9N3O2.H2O/c1-7(4(5)6)2-3(8)9;/h2H2,1H3,(H3,5,6)(H,8,9);1H2/i1D3; |
| SMILES | O.[2 H]C([2 H])([2 H])N(CC(=O)O)C(=N)N |
| IUPAC Name | 2-[carbamimidoyl(trideuteriomethyl)amino]acetic acid;hydrate |
| Molecular Weight (g/mol) | 152.167 |
| Formula Weight | 152.099 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction