missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine-d5 H2O (N-methyl-d3; glycine-2,2-d2), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Creatine-d5 Hydrate (N-methyl-d3; glycine-2,2-d2) |
| Molecular Formula | C4 H4 D5 N3 O2 . H2 O |
| Quantity | 0.1 g |
| Synonym | Creatine-d5 H2O (N-methyl-d3, glycine-2,2-d2) |
| InChI Formula | InChI=1 S/C4H9N3O2.H2O/c1-7(4(5)6)2-3(8)9;/h2H2,1H3,(H3,5,6)(H,8,9);1H2/i1D3,2D2; |
| SMILES | O.[2 H]C([2 H])([2 H])N(C(=N)N)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2-[carbamimidoyl(trideuteriomethyl)amino]-2,2-dideuterio-acetic acid;hydrate |
| Molecular Weight (g/mol) | 154.1792 |
| Formula Weight | 154.1114 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction