missing translation for 'onlineSavingsMsg'
Learn More

Creatine monohydrate, 99%

Product Code. 11431607
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431607 100 g 100g
11441607 500 g 500g
11451607 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431607 Supplier Thermo Scientific Alfa Aesar Supplier No. B25009.22
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6020-87-7 | C4H11N3O3 | 149.15 g/mol

Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6020-87-7
Molecular Formula C4H11N3O3
Molecular Weight (g/mol) 149.15
MDL Number MFCD00071582
InChI Key MEJYXFHCRXAUIL-UHFFFAOYSA-N
Synonym creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure
PubChem CID 80116
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=N)N.O

Specifications

Melting Point ∼292°C (decomposition)
pH 6.9
Odor Weak
Quantity 100 g
Beilstein 907175
Merck Index 14,2568
Solubility Information Soluble in water (17g/L) at 20°C. Insoluble in ether.
Formula Weight 149.15 (131.13 Anhydrous)
Percent Purity 99%
Chemical Name or Material Creatine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.