missing translation for 'onlineSavingsMsg'
Learn More

Creatine monohydrate, 99%

Product Code. 11441607
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11441607

missing translation for 'mfr': Thermo Scientific Alfa Aesar B25009.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6020-87-7 | C4H11N3O3 | 149.15 g/mol

Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.

Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6020-87-7
Molecular Formula C4H11N3O3
Molecular Weight (g/mol) 149.15
MDL Number MFCD00071582
InChI Key MEJYXFHCRXAUIL-UHFFFAOYSA-N
Synonym creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure
PubChem CID 80116
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=N)N.O

Specifications

Melting Point ∼292°C (decomposition)
pH 6.9
Odor Weak
Quantity 500 g
Beilstein 907175
Merck Index 14,2568
Solubility Information Soluble in water (17g/L) at 20°C. Insoluble in ether.
Formula Weight 149.15 (131.13 Anhydrous)
Percent Purity 99%
Chemical Name or Material Creatine monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.